3,5-Dichloro-2,4,6-trifluorobenzotrifluoride

Supplier: Matrix Scientific

007426-25G 007426-100G
101792-730EA 44.62 USD
101792-730 101792-732
3,5-Dichloro-2,4,6-trifluorobenzotrifluoride
3,5-Dichloro-2,4,6-trifluorobenzotrifluoride

Hazard: Irritant

Formula: C₇Cl₂F₆
Boiling Pt: 167-168°C/748mm
Density: 1.715
MDL Number: MFCD00039304
CAS Number: 4284-10-0

Order Now

Specification Test Results

Boiling point 167-168°C/7
Density 1.715
ND20 1.4456

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR