2-(Perfluorohexyl)acetaldehyde dimethyl acetal ≥97%

Supplier: Matrix Scientific

007116-5G 007116-25G
101792-056EA 112.31 USD
101792-056 101792-058
2-(Perfluorohexyl)acetaldehyde dimethyl acetal ≥97%
2-(Perfluorohexyl)acetaldehyde dimethyl acetal

Hazard: Irritant

Formula: C₁₀H₉F₁₃O₂
Boiling Pt: 71 °C (14 mmHg)
Density: 1.534
MDL Number: MFCD00236703
CAS Number: 142502-76-9

Order Now

Specification Test Results

Boiling point 71°C/14mm
Density 1.534
ND20 1.322


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR