(Perfluoro-n-decyl)ethane

Supplier: Matrix Scientific

007108-5G 007108-25G
101792-040EA 138.46 USD
101792-040 101792-042
(Perfluoro-n-decyl)ethane
(Perfluoro-n-decyl)ethane

Hazard: Irritant

Formula: C₁₂H₅F₂₁
Boiling Pt: 90°C/20mm
Density: 1.703
MDL Number: MFCD00236699
CAS Number: 154478-87-2

Order Now

Specification Test Results

Boiling point 90°C/20mm
Density 1.703
ND20 1.3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR