tert-Butoxydiphenylchlorosilane ≥97%

Supplier: Matrix Scientific

Synonyms: tert-Butoxychlorodiphenylsilane, tert-Butoxydiphenylsilyl Chloride

006721-1G 006721-5G
101790-964EA 23.08 USD
101790-964 101790-966
tert-Butoxydiphenylchlorosilane ≥97%
tert-Butoxydiphenylchlorosilane

Hazard: Flammable/Corrosive/Stored under Argon/Store in Freezer/Moisture Sensitive

Formula: C₁₆H₁₉ClOSi
MW: 290.86 g/mol
Boiling Pt: 130-135°C/0.1mm
Density: 1.057
MDL Number: MFCD00077672
CAS Number: 17922-24-6

Order Now

Specification Test Results

Boiling point 130-135°C/0
Density 1.057
ND20 1.538

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR