4-(Methylthio)toluene ≥99%

Supplier: Matrix Scientific

Synonyms: 4-Methylthioanisole, Methyl p-Tolyl sulphide

006348-25G 006348-100G
101790-402EA 538.46 USD
101790-402 101790-404
4-(Methylthio)toluene ≥99%
4-(Methylthio)toluene

TCA listed: T
Hazard: Irritant/Stench

Formula: C₈H₁₀S
MW: 138.23 g/mol
Boiling Pt: 52-54°C/1mm
Density: 1.027
MDL Number: MFCD00008560
CAS Number: 623-13-2

Order Now

Specification Test Results

Boiling point 52-54°C/1mm
Density 1.027
ND20 1.573

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR