3-Phenoxyphenylacetonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(3-Phenoxyphenyl)acetonitrile

005712-1G 005712-5G
101789-408EA 75.38 USD
101789-408 101789-410
3-Phenoxyphenylacetonitrile ≥98%
3-Phenoxyphenylacetonitrile

TCA listed: T
Hazard: Irritant

Formula: C₁₄H₁₁NO
Boiling Pt: 145°C/1mm
Density: 1.12
MDL Number: MFCD00016393
CAS Number: 51632-29-2
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 145°C/1mm
Density 1.12
ND20 1.578


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR