2-Methoxythioanisole ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(Methylsulphanyl)anisole

011172-5G 011172-25G
101800-068EA 73.85 USD
101800-068 101800-070
2-Methoxythioanisole ≥98%
2-Methoxythioanisole

Hazard: Irritant/Stench

Formula: C₈H₁₀OS
Boiling Pt: 96°C/16 Torr
Density: 1.12
MDL Number: MFCD00792424
CAS Number: 2388-73-0

Order Now

Specification Test Results

Boiling point 246°C
Density 1.12
ND20 1.595

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR