3-(Perfluorohexyl)propanol-1

Supplier: Matrix Scientific

Synonyms: 1H,1H,2H,2H,3H,3H-Tridecafluoro-1-nonanol, 3-(Perfluorohexyl)propanol

005292-5G 005292-25G
101788-600EA 141.54 USD
101788-600 101788-602
3-(Perfluorohexyl)propanol-1
3-(Perfluorohexyl)propanol-1

Hazard: Irritant

Formula: C₉H₇F₁₃O
Boiling Pt: 80 °C (10 mmHg)
Density: 1.629
MDL Number: MFCD00077578
CAS Number: 80806-68-4

Order Now

Specification Test Results

Boiling point 80°C/10mm
Density 1.629
ND20 1.329


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR