4'-(2-Chloroethyl)acetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-[4-(2-Chloroethyl)phenyl]ethan-1-one

005274-5G 005274-25G
101788-560EA 127.69 USD
101788-560 101788-562
4'-(2-Chloroethyl)acetophenone ≥98%
4'-(2-Chloroethyl)acetophenone

Hazard: Irritant

Formula: C₁₀H₁₁ClO
MW: 182.65
Boiling Pt: 112°C/0.1mm
Density: 1.15
MDL Number: MFCD00016350
CAS Number: 69614-95-5

Order Now

Specification Test Results

Boiling point 112°C/0.1mm
Density 1.15
ND20 1.555

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR