3-(Perfluorobutyl)-2-hydroxypropyl acrylate ≥97%

Supplier: Matrix Scientific

005257-5G 005257-25G
101788-544EA 581.54 USD
101788-544 101788-546
3-(Perfluorobutyl)-2-hydroxypropyl acrylate ≥97%
3-(Perfluorobutyl)-2-hydroxypropyl acrylate

Hazard: Irritant

Formula: C₁₀H₉F₉O₃
Boiling Pt: 89-90°C/3mm
Density: 1.442
MDL Number: MFCD00236105
CAS Number: 98573-25-2

Order Now

Specification Test Results

Boiling point 89-90°C/3mm
Density 1.442
ND20 1.353

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR