1-(Trifluoroacetyl)imidazole ≥95%

Supplier: Matrix Scientific

005142-5G 005142-25G 005142-100G
101788-322EA 30.77 USD
101788-322 101788-324 101788-326
1-(Trifluoroacetyl)imidazole ≥95%
1-(Trifluoroacetyl)imidazole

TCA listed: T
Hazard: Flammable/Moisture Sensitive/Hygroscopic/Stored under Argon/Store in Freezer

Formula: C₅H₃F₃N₂O
Boiling Pt: 45-46°C/14mm
Density: 1.442
Flash Pt: 45°C
MDL Number: MFCD00014501
CAS Number: 1546-79-8

Order Now

Specification Test Results

Boiling point 45-46°C/14m
Density 1.442
ND20 1.424


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR