3-(Perfluoro-n-hexyl)prop-2-en-1-ol ≥98%

Supplier: Matrix Scientific

005136-1G 005136-5G 005136-25G
101788-312EA 78.46 USD
101788-312 101788-314 101788-316
3-(Perfluoro-n-hexyl)prop-2-en-1-ol ≥98%
3-(Perfluoro-n-hexyl)prop-2-en-1-ol

Hazard: Irritant

Formula: C₉H₅F₁₃O
Boiling Pt: 87…89 °C (10 mmHg)
Density: 1.65
MDL Number: MFCD00042355
CAS Number: 38550-47-9

Order Now

Specification Test Results

Boiling point 87-89°C/10m
Density 1.65
ND20 1.332


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR