(Perfluoro-n-octyl)ethane ≥97%
Supplier: Matrix Scientific
004812-5G
004812-25G
101787-850EA
89.23
USD
101787-850
101787-852
(Perfluoro-n-octyl)ethane ≥97%
(Perfluoro-n-octyl)ethane
Hazard: Irritant
Formula:
C₁₀H₅F₁₇ Boiling Pt: 77°C/50mm Density: 1.654 |
MDL Number:
MFCD00236711 CAS Number: 77117-48-7 |
Specification Test Results
Boiling point | 77°C/50mm |
Density | 1.654 |
ND20 | 1.3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...