1,2-Dibromo-3-fluoropropane ≥98%

Supplier: Matrix Scientific

004548-1G 004548-5G
101787-452EA 53.85 USD
101787-452 101787-454
1,2-Dibromo-3-fluoropropane ≥98%
1,2-Dibromo-3-fluoropropane

TCA listed: T
Hazard: Irritant

Formula: C₃H₅Br₂F
Boiling Pt: 163.5°C
Density: 2.089
MDL Number: MFCD02093301
CAS Number: 453-00-9

Order Now

Specification Test Results

Boiling point 163.5°C
Density 2.089
ND20 1.5102

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR