1,3-Dibromo-1,1-difluorobutane ≥92%, Technical Grade
Supplier: Matrix Scientific
004531-5G
004531-25G
101787-404EA
103.08
USD
101787-404
101787-406
1,3-Dibromo-1,1-difluorobutane ≥92%, Technical Grade
1,3-Dibromo-1,1-difluorobutane
Hazard: Irritant
Formula:
C₄H₆Br₂F₂ Boiling Pt: 60-61°C/50mm Density: 1.8586 |
CAS Number:
406-42-8 |
Specification Test Results
Boiling point | 60-61°C/50m |
Density | 1.8586 |
ND20 | 1.4489 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...