Methyl thiosalicylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-mercaptobenzoate

004212-5G 004212-25G
101786-962EA 80 USD
101786-962 101786-964
Methyl thiosalicylate ≥97%
Methyl thiosalicylate

Hazard: Irritant

Formula: C₈H₈O₂S
MW: 168.21 g/mol
Boiling Pt: 98-100°C/2mm
Density: 1.22
MDL Number: MFCD00060678
CAS Number: 4892-02-8

Order Now

Specification Test Results

Boiling point 98-100°C/2m
Density 1.22
ND20 1.591

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR