2,2,2-Trifluoroethyl butyrate ≥98%

Supplier: Matrix Scientific

Synonyms: Butyric acid 2,2,2-Trifluoroethyl ester

003709-5G 003709-25G
101786-108EA 69.23 USD
101786-108 101786-110
2,2,2-Trifluoroethyl butyrate ≥98%
2,2,2-Trifluoroethyl butyrate

Hazard: Irritant

Formula: C₆H₉F₃O₂
Boiling Pt: 113°C/747mm
Density: 1.127
MDL Number: MFCD00077609
CAS Number: 371-27-7

Order Now

Specification Test Results

Boiling point 113°C/747mm
Density 1.127
ND20 1.347


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR