Diethyl mesoxalate (diethyl ketomalonate) ≥95%
Supplier: Matrix Scientific
Synonyms:
Diethyl 2-oxomalonate
003422-25G
003422-100G
101785-686EA
60
USD
101785-686
101785-688
Diethyl mesoxalate (diethyl ketomalonate) ≥95%
Diethyl mesoxalate (diethyl ketomalonate)
TCA listed: T
Hazard: Irritant/Moisture Sensitive
Formula:
C₇H₁₀O₅ MW: 174.15 g/mol Boiling Pt: 208…210 °C Density: 1.142 |
MDL Number:
MFCD00009121 CAS Number: 609-09-6 |
Specification Test Results
Boiling point | 208-210°C |
Density | 1.142 |
ND20 | 1.415 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...