2-Ethoxybenzonitrile ≥98%

Supplier: Matrix Scientific

003296-5G 003296-25G
101785-540EA 26.15 USD
101785-540 101785-542
2-Ethoxybenzonitrile ≥98%
2-Ethoxybenzonitrile

Hazard: Toxic

Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 154 °C (20 mmHg)
Density: 1.053
MDL Number: MFCD00001789
CAS Number: 6609-57-0

Order Now

Specification Test Results

Boiling point 154°C/20mm
Density 1.053
ND20 1.5322


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR