Methyl phenoxyacetate ≥99%

Supplier: Matrix Scientific

003189-25G 003189-100G
101785-382EA 18.46 USD
101785-382 101785-384
Methyl phenoxyacetate ≥99%
Methyl phenoxyacetate

TCA listed: T
Hazard: Irritant

Formula: C₉H₁₀O₃
Boiling Pt: 243 °C
Density: 1.149
MDL Number: MFCD00010227
CAS Number: 2065-23-8

Order Now

Specification Test Results

Boiling point 243°C
Density 1.149
ND20 1.514


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR