2,2,3-Trichloro-1,1,1,4,4,4-hexafluorobutane ≥97%

Supplier: Matrix Scientific

Synonyms: 1,1,1,4,4,4-Hexafluoro-2,2,3-trichlorobutane

003078-1G 003078-5G
101785-166EA 135.38 USD
101785-166 101785-168
2,2,3-Trichloro-1,1,1,4,4,4-hexafluorobutane ≥97%
2,2,3-Trichloro-1,1,1,4,4,4-hexafluorobutane

Hazard: Irritant

Formula: C₄HCl₃F₆
MW: 269.4 g/mol
Boiling Pt: 105 °C
Density: 1.69
MDL Number: MFCD02093295
CAS Number: 378-84-7

Order Now

Specification Test Results

Boiling point 105°C
Density 1.69
ND20 1.3645

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR