1-Bromo-2,5-bis(trifluoromethyl)benzene ≥97%

Supplier: Matrix Scientific

Synonyms: 2,5-Bis(trifluoromethyl)bromobenzene, 2-Bromo-1,4-bis(trifluoromethyl)benzene

009721-5G 009721-25G
101797-162EA 21.54 USD
101797-162 101797-164
1-Bromo-2,5-bis(trifluoromethyl)benzene ≥97%
1-Bromo-2,5-bis(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₈H₃BrF₆
MW: 293.01 g/mol
Boiling Pt: 146-147°C
Melting Pt: 4-5°C
Density: 1.743
MDL Number: MFCD03094336
CAS Number: 7617-93-8

Order Now

Specification Test Results

Boiling point 146-147°C
Density 1.691
ND20 1.434

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR