3-(2-Pyrrolidinyl)pyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(3-Pyridyl)pyrrolidine, (±)-Nornicotine, DL-Nornicotine, 3-(Pyrrolidin-2-yl)pyridine
009430-1G
009430-5G
101797-084EA
120
USD
101797-084
101797-086
3-(2-Pyrrolidinyl)pyridine ≥95%
3-(2-Pyrrolidinyl)pyridine
Hazard: Irritant/Irritant-Harmful/Refrigerate
Formula:
C₉H₁₂N₂ MW: 148.21 g/mol Boiling Pt: 107-108°C/2mm Density: 1.074 |
MDL Number:
MFCD00016913 CAS Number: 5746-86-1 |
Specification Test Results
Boiling point | 107-108°C/2 |
Density | 1.074 |
ND20 | 1.54 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...