1-[3-(Trifluoromethyl)phenyl]propan-1-ol ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(3-(Trifluoromethyl)phenyl)propan-1-ol

009209-1G 009209-5G
101796-642EA 121.54 USD
101796-642 101796-644
1-[3-(Trifluoromethyl)phenyl]propan-1-ol ≥97%
1-[3-(Trifluoromethyl)phenyl]propan-1-ol

Hazard: Irritant

Formula: C₁₀H₁₁F₃O
MW: 204.19 g/mol
Boiling Pt: 88…91 °C (5 mmHg)
Density: 1.197
MDL Number: MFCD00046668
CAS Number: 618-97-3

Order Now

Specification Test Results

Boiling point 88-91°C/5mm
Density 1.197
ND20 1.4582

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR