11-(Perfluoro-n-octyl)undec-10-en-1-ol ≥97%

Supplier: Matrix Scientific

009131-1G 009131-5G
101796-440EA 86.15 USD
101796-440 101796-442
11-(Perfluoro-n-octyl)undec-10-en-1-ol ≥97%
11-(Perfluoro-n-octyl)undec-10-en-1-ol

Hazard: Irritant

Formula: C₁₉H₂₁F₁₇O
Boiling Pt: 157 °C (3 mmHg)
Density: 1.417
MDL Number: MFCD00236713
CAS Number: 15364-19-9

Order Now

Specification Test Results

Boiling point 157°C/3mm
Density 1.417
ND20 1.37

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR