Dimethylpentafluorophenylsilane ≥98%

Supplier: Matrix Scientific

008931-5G 008931-25G
101795-842EA 483.08 USD
101795-842 101795-844
Dimethylpentafluorophenylsilane ≥98%
Dimethylpentafluorophenylsilane

Hazard: Flammable/Moisture Sensitive

Formula: C₈H₇F₅Si
Boiling Pt: 61…62 °C (20 mmHg)
Density: 1.295
Flash Pt: 40 °C
MDL Number: MFCD00010661
CAS Number: 13888-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR