Allyl-2-chloro-1,1,2-trifluoroethyl ether ≥97%
Supplier: Matrix Scientific
008788-5G
008788-25G
101795-460EA
100
USD
101795-460
101795-462
Allyl-2-chloro-1,1,2-trifluoroethyl ether ≥97%
Allyl-2-chloro-1,1,2-trifluoroethyl ether
Hazard: Irritant
Formula:
C₅H₆ClF₃O MW: 174.55 g/mol Boiling Pt: 109 °C (750 mmHg) Density: 1.25 |
MDL Number:
MFCD00039336 CAS Number: 380-44-9 |
Specification Test Results
Boiling point | 109°C/750mm |
Density | 1.25 |
ND20 | 1.3694 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...