1H,1H,5H-Octafluoropentyl methacrylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methacrylic Acid 1H,1H,5H-Perfluoropentyl Ester, Methacrylic Acid 1H,1H,5H-Octafluoropentyl Ester, 1H,1H,5H-Perfluoropentyl Methacrylate, 2,2,3,3,4,4,5,5-Octafluoropentyl methacrylate

002860-5G 002860-25G 002860-100G
101784-778EA 29.23 USD
101784-778 101784-780 101784-782
1H,1H,5H-Octafluoropentyl methacrylate ≥97%
1H,1H,5H-Octafluoropentyl methacrylate

Hazard: Irritant/Refrigerate

Formula: C₉H₈F₈O₂
MW: 300.15 g/mol
Boiling Pt: 80°C/23mm
Density: 1.5
Flash Pt: 81°C
MDL Number: MFCD00039278
CAS Number: 355-93-1

Order Now

Specification Test Results

Boiling point 80°C/23mm
Density 1.5
ND20 1.355

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR