1H,1H,2H,2H-Tridecafluoro-n-octyl acrylate ≥95%

Supplier: Matrix Scientific

Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl acrylate, Acrylic acid 1H,1H,2H,2H-Tridecafluoro-n-octyl ester

002232-5G 002232-25G
101783-502EA 66.15 USD
101783-502 101783-504
1H,1H,2H,2H-Tridecafluoro-n-octyl acrylate ≥95%
1H,1H,2H,2H-Tridecafluoro-n-octyl acrylate

TCA listed: T
Hazard: Irritant/Refrigerate

Formula: C₁₁H₇F₁₃O₂
MW: 418.154 g/mol
Boiling Pt: 76-80°C/8mm
Density: 1.554
MDL Number: MFCD00042351
CAS Number: 17527-29-6

Order Now

Specification Test Results

Boiling point 76-80°C/8mm
Density 1.554
ND20 1.338

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR