Benzyl-2,2,2-trifluoroethyl sulfide ≥97%

Supplier: Matrix Scientific

001935-1G 001935-5G
101782-882EA 38.46 USD
101782-882 101782-884
Benzyl-2,2,2-trifluoroethyl sulfide ≥97%
Benzyl-2,2,2-trifluoroethyl sulfide

Hazard: Flammable

Formula: C₉H₉F₃S
MW: 206.23 g/mol
Boiling Pt: 45…46 °C (20 mmHg)
Density: 1.222
Flash Pt: 10 °C
MDL Number: MFCD00191786
CAS Number: 77745-03-0

Order Now

Specification Test Results

Boiling point 45-46°C/20m
Density 1.222
ND20 1.491

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR