Isoflurane ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether

001932-25G 001932-100G
101782-874EA 135.38 USD
101782-874 101782-876
Isoflurane ≥97%
Isoflurane

Hazard: Irritant/Refrigerate

Formula: C₃H₂ClF₅O
MW: 184.49 g/mol
Boiling Pt: 49°C
Density: 1.45
MDL Number: MFCD00066609
CAS Number: 26675-46-7

Order Now

Specification Test Results

Boiling point 49°C
Density 1.45
ND20 1.3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR