2-Chloro-1,1,2-trifluoroethyl difluoromethyl ether ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-1H,4H-perfluoro(2-oxabutane)

001888-25G 001888-100G
101782-810EA 227.69 USD
101782-810 101782-812
2-Chloro-1,1,2-trifluoroethyl difluoromethyl ether ≥97%
2-Chloro-1,1,2-trifluoroethyl difluoromethyl ether

Hazard: Irritant

Formula: C₃H₂ClF₅O
MW: 184.49 g/mol
Boiling Pt: 56 °C
Density: 1.517
MDL Number: MFCD00069095
CAS Number: 13838-16-9

Order Now

Specification Test Results

Boiling point 56°C
Density 1.517
ND20 1.303

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR