2,2,3,3,3-Pentafluoropropyl-1,1,2,2-tetrafluoroethyl ether ≥97%

Supplier: Matrix Scientific

001570-5G 001570-25G
101781-982EA 300 USD
101781-982 101781-984
2,2,3,3,3-Pentafluoropropyl-1,1,2,2-tetrafluoroethyl ether ≥97%
2,2,3,3,3-Pentafluoropropyl-1,1,2,2-tetrafluoroethyl ether

Hazard: Irritant

Formula: C₅H₃F₉O
Boiling Pt: 68 °C
Density: 1.567
MDL Number: MFCD00236117
CAS Number: 50807-74-4

Order Now

Specification Test Results

Boiling point 68°C
Density 1.567
ND20 1.283

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR