Methyl-2,2,3,3,3-pentafluoropropyl ether ≥97%

Supplier: Matrix Scientific

001566-5G 001566-25G
101781-974EA 384.62 USD
101781-974 101781-976
Methyl-2,2,3,3,3-pentafluoropropyl ether ≥97%
Methyl-2,2,3,3,3-pentafluoropropyl ether

Hazard: Irritant

Formula: C₄H₅F₅O
MW: 164.08 g/mol
Boiling Pt: 46°C
Density: 1.269
MDL Number: MFCD00236115
CAS Number: 378-16-5

Order Now

Specification Test Results

Boiling point 46°C
Density 1.269
ND20 1.285

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR