1H,1H,3H-Hexafluorobutyl acrylate

Supplier: Matrix Scientific

Synonyms: HFBA, 2,2,3,4,4,4-Hexafluorobutyl Acrylate, Acrylic acid 2,2,3,4,4,4-Hexafluorobutyl ester

001233-5G 001233-25G
101781-220EA 84.62 USD
101781-220 101781-222
1H,1H,3H-Hexafluorobutyl acrylate
1H,1H,3H-Hexafluorobutyl acrylate

Hazard: Irritant/Refrigerate/Lachrymator

Formula: C₇H₆F₆O₂
MW: 236.1118 g/mol
Boiling Pt: 40…43 °C (8 mmHg)
Density: 1.389
MDL Number: MFCD00044091
CAS Number: 54052-90-3

Order Now

Specification Test Results

Boiling point 40-43°C/8mm
Density 1.389
ND20 1.352


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR