1,1,2,3,3,3-Hexafluoropropyl methyl ether ≥97%

Supplier: Matrix Scientific

001191-5G 001191-25G
101781-116EA 153.85 USD
101781-116 101781-118
1,1,2,3,3,3-Hexafluoropropyl methyl ether ≥97%
1,1,2,3,3,3-Hexafluoropropyl methyl ether

Hazard: Irritant

Formula: C₄H₄F₆O
Boiling Pt: 53 °C
Density: 1.419
MDL Number: MFCD00041537
CAS Number: 382-34-3

Order Now

Specification Test Results

Boiling point 53°C
Density 1.419
ND20 1.284

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR