3-Bromo-3,3-difluoropropene ≥98%

Supplier: Matrix Scientific

Synonyms: 3,3-Difluoro-3-bromopropene

001133-1G 001133-5G
101781-020EA 223.08 USD
101781-020 101781-022
3-Bromo-3,3-difluoropropene ≥98%
3-Bromo-3,3-difluoropropene

Hazard: Irritant/Refrigerate

Formula: C₃H₃BrF₂
MW: 156.96 g/mol
Boiling Pt: 42°C
Density: 1.543
MDL Number: MFCD00042185
CAS Number: 420-90-6

Order Now

Specification Test Results

Boiling point 42°C
Density 1.543
ND20 1.377

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR