4-Bromo-2,5-dimethylanisole ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-4-methoxy-2,5-dimethylbenzene

056912-5G 056912-25G
101911-698EA 75.38 USD
101911-698 101911-700
4-Bromo-2,5-dimethylanisole ≥98%
4-Bromo-2,5-dimethylanisole

Hazard: Irritant

Formula: C₉H₁₁BrO
MW: 215.1 g/mol
Boiling Pt: 117 °C (5 mmHg)
Melting Pt: 34…36 °C
MDL Number: MFCD01652198
CAS Number: 58106-25-5

Order Now

Specification Test Results

Boiling point 117°C/5mm
Melting point 34-36°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR