Methyl-L-2-isothiocyanato-3-phenylpropionate ≥99%

Supplier: Matrix Scientific

056603-1G 056603-5G
101911-014EA 152.31 USD
101911-014 101911-016
Methyl-L-2-isothiocyanato-3-phenylpropionate ≥99%
Methyl-L-2-isothiocyanato-3-phenylpropionate

Hazard: Irritant-Harmful

Formula: C₁₁H₁₁NO₂S
Boiling Pt: 125 °C (1 mmHg)
Melting Pt: 34…36 °C
MDL Number: MFCD09025713
CAS Number: 68521-58-4

Order Now

Specification Test Results

Boiling point 125°C/1mm
Melting point 34-36°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR