4-Methoxybenzoyl isothiocyanate ≥98%
Supplier: Matrix Scientific
056574-1G
056574-5G
101910-924EA
107.69
USD
101910-924
101910-926
4-Methoxybenzoyl isothiocyanate ≥98%
4-Methoxybenzoyl isothiocyanate
Hazard: Irritant-Harmful
Formula:
C₉H₇NO₂S Boiling Pt: 122 °C (1 mmHg) Melting Pt: 23…25 °C |
MDL Number:
MFCD00060379 CAS Number: 16778-84-0 |
Specification Test Results
Boiling point | 122°C/1mm |
Melting point | 23-25°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...