2,5-Dichloro-3-(trifluoromethyl)pyridine ≥97%
Supplier: Matrix Scientific
048523-500MG
048523-1G
048523-5G
101884-202EA
153.85
USD
101884-202
101884-204
101884-206
2,5-Dichloro-3-(trifluoromethyl)pyridine ≥97%
2,5-Dichloro-3-(trifluoromethyl)pyridine
Hazard: Irritant
Formula:
C₆H₂Cl₂F₃N MW: 215.9880 g/mol Boiling Pt: 80…82 °C (26 mmHg) |
MDL Number:
MFCD00042244 CAS Number: 70158-59-7 |
Specification Test Results
Boiling point | 80-82/26mm |
Melting point | 80-82/26mm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...