Diethyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]malonate ≥95%

Supplier: Matrix Scientific

045820-500MG 045820-1G 045820-5G
101872-778EA 375.38 USD
101872-778 101872-780 101872-782
Diethyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]malonate ≥95%
Diethyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]malonate

Hazard: Irritant

Formula: C₁₃H₁₃ClF₃NO₄
MW: 339.7 g/mol
Boiling Pt: 120…124 °C (0.5 mmHg)
Melting Pt: 43…45 °C
MDL Number: MFCD00231883
CAS Number: 172527-71-8

Order Now

Specification Test Results

Boiling point 120-124°C/0
Melting point 43-45°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR