2-(4-Chlorophenyl)-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]acetonitrile ≥95%

Supplier: Matrix Scientific

044077-500MG 044077-1G 044077-5G
101864-008EA 375.38 USD
101864-008 101864-010 101864-012
2-(4-Chlorophenyl)-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]acetonitrile ≥95%
2-(4-Chlorophenyl)-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]acetonitrile

Hazard: Irritant

Formula: C₁₄H₇Cl₂F₃N₂
MW: 331.13 g/mol
Boiling Pt: 146°C/0.1mm
Melting Pt: 47…49 °C
MDL Number: MFCD00172211
CAS Number: 213993-80-7

Order Now

Specification Test Results

Boiling point 146°C/0.1mm
Melting point 47-49°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR