2-(3,4-Difluorophenoxy)benzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(3,4-Difluorophenoxy)benzenecarboxaldehyde

043521-500MG 043521-1G 043521-5G
101861-240EA 310.77 USD
101861-240 101861-242 101861-244
2-(3,4-Difluorophenoxy)benzaldehyde ≥95%
2-(3,4-Difluorophenoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₃H₈F₂O₂
MW: 234.1982 g/mol
Boiling Pt: 110…112 °C (0.2 mmHg)
Melting Pt: 61…64 °C
MDL Number: MFCD01568793
CAS Number: 289717-89-1

Order Now

Specification Test Results

Boiling point 110-112°C/0
Melting point 61-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR