2-(4-Fluorophenoxy)benzaldehyde ≥95%

Supplier: Matrix Scientific

043260-500MG 043260-1G 043260-5G
101859-944EA 153.85 USD
101859-944 101859-946 101859-948
2-(4-Fluorophenoxy)benzaldehyde ≥95%
2-(4-Fluorophenoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₃H₉FO₂
MW: 216.21 g/mol
Boiling Pt: 120-122°C/0.05mm
Melting Pt: 54-56°C
MDL Number: MFCD01568770
CAS Number: 320423-61-8

Order Now

Specification Test Results

Boiling point 120-122°C/0
Melting point 54-56°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR