2-(3-Chlorophenoxy)benzaldehyde ≥95%

Supplier: Matrix Scientific

043259-500MG 043259-1G 043259-5G
101859-938EA 424.44 USD
101859-938 101859-940 101859-942
2-(3-Chlorophenoxy)benzaldehyde ≥95%
2-(3-Chlorophenoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₃H₉ClO₂
Boiling Pt: 130-132°C/0.05mm
Melting Pt: 35-37°C
MDL Number: MFCD01568769
CAS Number: 181297-72-3

Order Now

Specification Test Results

Boiling point 130-132°C/0
Melting point 35-37°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR