2-(2,4-Dichlorophenoxy)benzaldehyde ≥95%

Supplier: Matrix Scientific

043258-500MG 043258-1G
101859-934EA 747.69 USD
101859-934 101859-936
2-(2,4-Dichlorophenoxy)benzaldehyde ≥95%
4-(2,4-Dichlorophenoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₃H₈Cl₂O₂
Boiling Pt: 147°C/0.1mm
Melting Pt: 58-60°C
MDL Number: MFCD01568767
CAS Number: 86309-06-0

Order Now

Specification Test Results

Boiling point 147°C/0.1mm
Melting point 58-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR