2-(Dimethylamino)-2-(1-methyl-1H-pyrrol-2-yl)acetonitrile ≥95%
Supplier: Matrix Scientific
042534-500MG
042534-1G
042534-5G
101856-232EA
375.38
USD
101856-232
101856-234
101856-236
2-(Dimethylamino)-2-(1-methyl-1H-pyrrol-2-yl)acetonitrile ≥95%
2-(Dimethylamino)-2-(1-methyl-1H-pyrrol-2-yl)acetonitrile
Hazard: Irritant
Formula:
C₉H₁₃N₃ Boiling Pt: 110°C/5mm Melting Pt: 39…41 °C |
MDL Number:
MFCD00231710 CAS Number: 117068-07-2 |
Specification Test Results
Boiling point | 110°C/5mm |
Melting point | 39-41°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...