[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-4-(hydroxymethyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole

042524-500MG 042524-1G 042524-5G
101856-178EA 556.92 USD
101856-178 101856-180 101856-182
[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol ≥95%
[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol

Hazard: Irritant

Formula: C₆H₆ClF₃N₂O
MW: 214.58 g/mol
Boiling Pt: 94…95 °C (0.8 mmHg)
Melting Pt: 94…95 °C
MDL Number: MFCD00141467
CAS Number: 282523-11-9

Order Now

Specification Test Results

Boiling point 94-95°C/0.8
Melting point 94-95°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR