Pyrimidine-5-carboxaldehyde

Supplier: Matrix Scientific

Synonyms: 5-Pyrimidinecarboxaldehyde

007321-1G 007321-5G
101792-446EA 38.46 USD
101792-446 102550-896
Pyrimidine-5-carboxaldehyde
Pyrimidine-5-carboxaldehyde

Hazard: Irritant/Store in Freezer

Formula: C₅H₄N₂O
MW: 108.1 g/mol
Boiling Pt: 80-82°C/4mm
Melting Pt: 39…43 °C
MDL Number: MFCD03426065
CAS Number: 10070-92-5

Order Now

Specification Test Results

Boiling point 80-82°C/4mm
Melting point 39-43°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR